E-Gel™ Sample Loading Buffer, 1X
Invitrogen™

E-Gel™ Sample Loading Buffer, 1X

E-Gel Sample Loading Buffer is supplied as a ready-to-use 1X solution, and is formulated specifically for maximum performance on E-GelRead more
Have Questions?
Catalog NumberQuantity
104820554 x 1.25 mL
Catalog number 10482055
Price (USD)
81.65
Online exclusive
85.00
Save 3.35
Each
Quantity:
4 x 1.25 mL
Price (USD)
81.65
Online exclusive
85.00
Save 3.35
Each
E-Gel Sample Loading Buffer is supplied as a ready-to-use 1X solution, and is formulated specifically for maximum performance on E-Gel EX gels as well as other E-Gel precast gels.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration1X
FormLiquid
Gel CompatibilityE-Gel™ agarose gels
Quantity4 x 1.25 mL
Sample Loading Volume1.25 mL
Shelf Life6 Months
Shipping ConditionApproved for shipment at Room Temperature or on Dry Ice
Gel TypeAgarose
Label or DyeTartrazine, Xylene Cyanol, Xylene Cyanol FF
Product LineE-Gel
Product TypeSample Loading Buffer
Unit SizeEach
Contents & Storage
• 4 x 1.25 mL 1X E-Gel Sample Loading buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.