Product Image
Thermo Scientific™

EpiSerf Medium

Gibco™ EpiSerf Medium is a serum-free medium for the culture of a wide variety of adherent cells, including primary epithelialRead more
Have Questions?
Catalog NumberQuantity
10732022500 mL
Catalog number 10732022
Price (USD)
146.00
Each
Quantity:
500 mL
Price (USD)
146.00
Each
Gibco™ EpiSerf Medium is a serum-free medium for the culture of a wide variety of adherent cells, including primary epithelial cells, MDBK, MDCK, VERO, and CHO. The growth and morphology is comparable to the same cells grown in a classic medium supplemented with Fetal Bovine Serum (FBS). Gibco™ Episerf Medium is a confidential formulation containing L-glutamine and 2 g/L total protein.

Product Use
For Research Use Only. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System
Gibco™ EpiSerf Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Specifications
Cell LineCHO, VERO, MDCK, MDBK
Cell TypeEpithelial Cells
Concentration1 X
Endotoxin Level≤10 EU/mL
Product TypeSerum Free Medium (SFM)
Quantity500 mL
Shelf Life12 Months
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Unit SizeEach
Contents & Storage
Storage conditions:2°C to 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture