UltraPure™ DEPC-treated Water, 4 x 1.25 mL - FAQs
UltraPure™ DEPC-treated Water, 4 x 1.25 mL - FAQs
View additional product information for UltraPure™ DEPC-Treated Water - FAQs (750023, 10813012, 750024)
1 product FAQs found
What is the shelf life of UltraPure DEPC-Treated Water?
The UltraPure DEPC-Treated Water is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.