Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
Human Endothelial SFM
Gibco™

Human Endothelial SFM

Human Endothelial Serum Free Medium (SFM) has been developed to support the long-term propagation of human umbilical vein endothelial cells (HUVEC).
Have Questions?
Catalog NumberQuantity
11111044500 mL
Catalog number 11111044
Price (USD)
163.00
Each
Quantity:
500 mL
Price (USD)
163.00
Each

Human Endothelial Serum Free Medium (SFM) has been developed to support the long-term propagation of human umbilical vein endothelial cells (HUVEC). Complete Human Endothelial SFM has been demonstrated to successfully support primary isolation and subsequent secondary growth of HUVEC for ≤15 passages. HUVEC cultured in complete Human Endothelial SFM exhibit the histotypic 'cobblestone' morphology and retain endothelial specific markers including: expression of Factor VIII-related antigen, UEA-1 lectin binding, uptake of DiI-acetylated LDL, vimentin, and IL-1a-induced ICAM-1 expression. Additionally, HUVEC cultured in Human Endothelial SFM have been shown to maintain cAMP and prostacyclin signal transduction systems. Human Endothelial SFM also supports the growth and retention of physiological markers for human umbilical arterial and dermal microvascular endothelial cells.

Human Endothelial SFM is a basal serum-free growth medium and requires supplementation with bFGF (20 ng/mL), EGF (10 ng/mL), and human plasma fibronectin (10 μg/mL).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineHUAEC, HUVEC
Cell TypeEndothelial Cells
Endotoxin Level≤10 EU/mL, ≤ 10 EU/mL
Product TypeSerum Free Medium (SFM)
Quantity500 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
ClassificationcGMP, Serum-free, Chemically-defined
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo Antibiotics
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

Do you offer a phenol red-free option for Human Endothelial-SFM (Cat. No. 11111044)?

We do not offer a phenol red-free version of this media as a standard catalog product. However, our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an “official” order or declining.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.