Search
View additional product information for Human Endothelial SFM - FAQs (11111044)
1 product FAQs found
We do not offer a phenol red-free version of this media as a standard catalog product. However, our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an official order or declining.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.