Sample and Skimmer Cones
Actual product may vary
Thermo Scientific™
Sample and Skimmer Cones
Choose from this range of sample and skimmer cones to suit your analysis requirements when using the Thermo Scientific™ ELEMENT™ Series of HR-ICP-MS instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
1142160Skimmer ConeNickel, X-Version
1044530Sample ConeNickel
1047460Skimmer ConePlatinum, Boron-free
1047461Skimmer ConePlatinum
1067500Sample ConePlatinum, Boron-free
1067501Sample ConePlatinum
1067600Skimmer ConeNickel
1142150Skimmer ConePlatinum, X-Vers, Boron-free
1142151Skimmer ConePlatinum, X-Version
1184050Sample ConeAluminum
1184060Skimmer ConeAluminum
1260630Jet Sample ConeNickel
BRE0010791Jet Sample ConePlatinum, Boron-free
BRE0010792Jet Sample ConePlatinum w/ Boron
Catalog number 1142160
Price (USD)
-
Type:
Skimmer Cone
Material:
Nickel, X-Version
A range of easily changeable Ni, Pt, and Al Thermo Scientific™ Sample and Skimmer Cones are available for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS and Thermo Scientific™ Neptune™ Series MC-ICP-MS systems to suit your analysis requirements, all designed for durability, performance and ease of use.
Select High-Quality Consumables for Consistent Performance
These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in solid Ni construction, Pt-tipped for even greater durability, and Al for specific analysis requirements.
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System and Neptune Series MC-ICP-MS System
MaterialNickel, X-Version
TypeSkimmer Cone
Unit SizeEach