PCR<sub>X</sub> Enhancer System
Invitrogen™

PCRX Enhancer System

PCRx Enhancer System is used in conjunction with DNA polymerases to optimize PCR of problematic and/or GC-rich templates.
Have Questions?
Catalog NumberQuantity
11495017250 Reactions
Catalog number 11495017
Price (USD)
112.65
Online exclusive
125.00
Save 12.35
Each
Quantity:
250 Reactions
Price (USD)
112.65
Online exclusive
125.00
Save 12.35
Each
PCRX Enhancer System is an optimized buffer and cosolvent system designed for use in conjunction with DNA polymerases to optimize PCR of problematic and/or GC-rich templates. 10X PCRX Enhancer Solution with 10X PCRX Amplification Buffer offers higher primer specificity, broader magnesium concentration, broader annealing temperature, and improved thermostability of DNA polymerases such as Taq DNA polymerase and Platinum Taq DNA Polymerase.

Conventional PCR buffer components can be directly substituted with the 10X PCRX amplification buffer and 50 mM MgSO4 for a more robust amplification and improved product yield in some cases.

Notes

Optimal concentration of 10X PCRX Enhancer Solution will vary depending on GC content, Mg++ concentration, and annealing temperature.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes• 10X PCRx Enhancer Solution (2 x 1 mL)
• 10X PCRx Amplification Buffer (2 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -20°C.
For Use With (Application)Standard PCR
Quantity250 Reactions
Unit SizeEach