Product Image
Invitrogen™

Platinum™ Quantitative PCR SuperMix-UDG w/ROX

**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’veRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
11743100100 Reactions
11743500500 Reactions
Catalog number 11743100
Price (USD)
499.65
Online exclusive
520.00
Save 20.35
Each
No. of Reactions:
100 Reactions
Price (USD)
499.65
Online exclusive
520.00
Save 20.35
Each
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of Platinum Quantitative PCR SuperMix and added additional capabilities for your gene expression analysis.

**********************
qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA. ROX reference dye is included in the mix for maximum convenience. These qPCR SuperMixes include:


–Platinum™ Taq DNA Polymerase: activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification (Figure 1)

–Uracil-DNA Glycosylase (UDG): prevents carryover contamination in conjunction with dUTP (included) for improved specificity
–ROX reference dye premixed at optimal concentration for ABI instruments


qPCR SuperMixes with ROX are available in two formats:

–Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye (Figure 2)
–Platinum™ qPCR SuperMix-UDG with ROX offers superior performance with either fluorogenic primers (e.g., LUX™ Primers) or probe-based detection systems (e.g., TaqMan™)


Contents and Storage:
Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX includes a 2X SuperMix with SYBR™ Green I and ROX Reference Dyes, and 50 mM MgCl2. Platinum™ qPCR SuperMix-UDG with ROX includes a 2X SuperMix with ROX Reference Dye, and 50 mM MgCl2. Store all components at -20°C. Guaranteed stable for 6 months when properly stored.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System
No. of Reactions100 Reactions
PolymerasePlatinum Taq DNA Polymerase
Product LinePlatinum
Product TypeQuantitative PCR SuperMix-UDG
Quantity100 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionDry Ice
Sufficient For100 Reactions
Volume2.5 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• 2 × 1.25 ml Platinum™ Quantitative PCR SuperMix-UDG with ROX
• 1 ml Magnesium Chloride (50 mM)

Store at -20°C in the dark.