Product Image
Invitrogen™

Platinum™ SYBR™ Green qPCR SuperMix-UDG w/ROX

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
11744500500 Reactions
11744100100 Reactions
Catalog number 11744500
Price (USD)
1,776.65
Online exclusive
1,890.00
Save 113.35
Each
No. of Reactions:
500 Reactions
Price (USD)
1,776.65
Online exclusive
1,890.00
Save 113.35
Each
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of Platinum SYBR Green qPCR SuperMix-UDG and added additional capabilities for your gene expression analysis.

**********************
qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA. ROX reference dye is included in the mix for maximum convenience. These qPCR SuperMixes include:


–Platinum™ Taq DNA Polymerase: activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification (Figure 1)

–Uracil-DNA Glycosylase (UDG): prevents carryover contamination in conjunction with dUTP (included) for improved specificity
–ROX reference dye premixed at optimal concentration for ABI instruments


qPCR SuperMixes with ROX are available in two formats:

–Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye (Figure 2)
–Platinum™ qPCR SuperMix-UDG with ROX offers superior performance with either fluorogenic primers (e.g., LUX™ Primers) or probe-based detection systems (e.g., TaqMan™)


Contents and Storage:
Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX includes a 2X SuperMix with SYBR™ Green I and ROX Reference Dyes, and 50 mM MgCl2. Platinum™ qPCR SuperMix-UDG with ROX includes a 2X SuperMix with ROX Reference Dye, and 50 mM MgCl2. Store all components at -20°C. Guaranteed stable for 6 months when properly stored.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
No. of Reactions500 Reactions
PolymeraseTaq DNA Polymerase
Product LinePlatinum, SYBR
Product TypeReal Time PCR SYBR Master Mix
Quantity500 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionDry Ice
Sufficient For500 Reactions of 50 μL Each
Volume12.5 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains 2X Platinum SYBR Green qPCR SuperMix-UDG with ROX (12.5 mL) and 50 mM Magnesium Chloride (2 × 1 mL).

Contains sufficient reagents for 500 reactions of 50 μL each. Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

What is the difference in sensitivity between TaqMan chemistry vs. SYBR Green reagent chemistry?

Sensitivity can actually be equivalent when using TaqMan chemistry and SYBR Green reagent chemistry. It might seem that a TaqMan assay with fluorescent signal generated by a sequence-specific probe would always be more sensitive than a SYBR Green reagent assay, but a poorly designed TaqMan assay could theoretically be less specific than a well-designed SYBR Green reagent assay. However, the potential for detection of primer dimers and non-specific products using SYBR Green chemistry is more likely to result in loss of sensitivity when attempting to quantitate lower copy numbers.

For more information on Real-Time PCR chemistries, please refer to the following Application Notes, which you can find on our website through Technical Resources, or by entering the titles in the main Search field: “Real-Time PCR Vs. Traditional PCR”, “Essentials of Real Time PCR”, and “Selection of Reagents for Real-Time PCR”.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

What are the key differences between a TaqMan MGB probe and a TaqMan TAMRA dye-labeled probe?

The TaqMan MGB probes contain the following features:
1) A fluorescent reporter at the 5' end
2) A nonfluorescent quencher at the 3' end. Because the quencher does not fluoresce, the real-time instruments can measure the reporter dye contributions more precisely.
3) A minor groove binder at the 3' end. The minor groove binder increases the melting temperature (Tm) of probes, allowing the use of shorter probes.

In general, the TaqMan MGB probes exhibit great differences in Tm values between matched and mismatched probes, which provides more accurate allelic discrimination and makes for a more sensitive real-time assay. Mismatches between a probe and allele, or target, reduce the efficiency of probe hybridization in a measurable way, which is especially important in SNP Genotyping assays. Furthermore, AmpliTaq Gold DNA polymerase is more likely to displace the mismatched probe rather than cleave it to release reporter dye. More information about TaqMan MGB probes can be found in the User Bulletin entitled "Primer Express Version 1.5 and TaqMan MGB Probes for Allelic Discrimination." You can find a copy on our website by entering this title in the main search field.

When using SYBR Green chemistry on an Applied Biosystems Real-Time PCR instrument, how do I change settings to reflect that there is no TaqMan probe being used in the reaction?

Refer to the product manual for your instrument and software for specifics, but in general you will want to change the Quencher value to None.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

How can RNA standards be generated to perform absolute quantitation for RNA targets?

It is generally not possible to use DNA as a standard for absolute quantitation of RNA because there is no control for the efficiency of the reverse transcription step. Therefore, in-vitro transcribed RNA is commonly used to prepare standards for the absolute quantitation of RNA targets. This would involve the cloning of the target of interest into an in-vitro transcription plasmid, performing in-vitro transcription, then purifying the resulting cRNA so that the DNA plasmid cannot serve as a PCR template. Concentration is measured by A260 and converted to the number of copies using the molecular weight of the RNA.

Relative quantitation of gene expression methods require less up-front preparation and provide a fold-change value instead of an absolute quantity result. For many researchers, absolute quantities are not a necessary parameter to measure, and therefore relative quantitation is a much more attractive approach to studying gene expression via real-time PCR. For more information on relative quantitation of gene expression, please refer to our Technical Reference Library in the Technical Resources section of our website.

Citations & References (1)

Citations & References
Abstract
Induction of pluripotent stem cells from adult human fibroblasts by defined factors.
Authors:Takahashi K, Tanabe K, Ohnuki M, Narita M, Ichisaka T, Tomoda K, Yamanaka S,
Journal:Cell
PubMed ID:18035408
Successful reprogramming of differentiated human somatic cells into a pluripotent state would allow creation of patient- and disease-specific stem cells. We previously reported generation of induced pluripotent stem (iPS) cells, capable of germline transmission, from mouse somatic cells by transduction of four defined transcription factors. Here, we demonstrate the generation ... More