SuperScript™ VILO™ cDNA Synthesis Kit, 50 reactions - FAQs
SuperScript™ VILO™ cDNA Synthesis Kit, 50 reactions - FAQs
View additional product information for SuperScript™ VILO™ cDNA Synthesis Kit - FAQs (11754050, 11754250)
5 product FAQs found
What is the difference between SuperScript III RT and the RT in the SuperScript VILO kit?
The SuperScript VILO cDNA Synthesis Kit contains a mix of SuperScript III RT and helper proteins which help to increase the efficiency of the reverse transcription reaction and thus improve yield. The RT in the SuperScript VILO kit is active at 42 degrees C due to the helper proteins.
How much of the first-strand cDNA reaction should I load for PCR?
While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.
In what concentration range of total RNA does the SuperScript VILO cDNA Synthesis Kit (Cat. No. 11754050) work?
  • Lower limit of RNA concentration: 1.0 pg
  • Upper limit of RNA concentration: 2.5 µg
  • Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center
    What is the RNA input for the Oncomine Focus Assay, Chef-Ready Library kit?
    Each reverse transcription reaction requires 10 ng of DNase-treated total RNA (≥1.43 ng/µL).
    With the Oncomine Focus Assay, Chef-Ready Library, can I perform reverse transcription using the SuperScript VILO cDNA Synthesis Kit on the Ion Chef System?
    Reverse transcription needs to be performed manually in the IonCode 96-well PCR plate, using the SuperScript VILO cDNA Synthesis Kit. SuperScript reverse transcriptase enzyme is not supplied in the kit and must be purchased separately. If the RNA was prepared from FFPE tissue and not previously heat-treated, we recommend heating at 80 degrees C for 10 min, then cooling to room temperature.