Product Image
Invitrogen™

SYBR GreenER™ qPCR SuperMix for iCycler™ Instrument

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
11761100100 Reactions
11761500500 Reactions
1176102K2000 Reactions
Catalog number 11761100
Price (USD)
400.00
Each
No. of Reactions:
100 Reactions
Price (USD)
400.00
Each
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of SYBR GreenER qPCR SuperMix and added additional capabilities for your gene expression analysis.

**********************
SYBR™ GreenER™ qPCR SuperMixes for iCycler™ incorporate the latest high-performance technology to provide the most reliable gene expression data from your iCycler iQ™ or MyiQ™ instrument. The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility with:
• A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
• An optimized buffer system - improves sensitivity and provides excellent long-term stability
• Uracil DNA glycosylase - reduces carryover contamination in qPCR

Novel Formulation Provides More Reliable Data
The SYBR™ GreenER™ qPCR SuperMix kits include a new dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.

Consistent Specificity and Low-Copy Detection
The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
No. of Reactions100 Reactions
PolymeraseTaq DNA Polymerase
Product LinePlatinum, SYBR GreenER
Product TypeReal Time PCR SYBR Master Mix
Quantity100 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionDry Ice
Sufficient For100 Reactions
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
2X SYBR™ GreenER™ qPCR SuperMix for iCycler™ instrument contains: hot-start Taq DNA polymerase,
SYBR™ GreenER™ fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers.

Store at 4°C upon receipt. Guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

What is the difference in sensitivity between TaqMan chemistry vs. SYBR Green reagent chemistry?

Sensitivity can actually be equivalent when using TaqMan chemistry and SYBR Green reagent chemistry. It might seem that a TaqMan assay with fluorescent signal generated by a sequence-specific probe would always be more sensitive than a SYBR Green reagent assay, but a poorly designed TaqMan assay could theoretically be less specific than a well-designed SYBR Green reagent assay. However, the potential for detection of primer dimers and non-specific products using SYBR Green chemistry is more likely to result in loss of sensitivity when attempting to quantitate lower copy numbers.

For more information on Real-Time PCR chemistries, please refer to the following Application Notes, which you can find on our website through Technical Resources, or by entering the titles in the main Search field: “Real-Time PCR Vs. Traditional PCR”, “Essentials of Real Time PCR”, and “Selection of Reagents for Real-Time PCR”.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

What are the key differences between a TaqMan MGB probe and a TaqMan TAMRA dye-labeled probe?

The TaqMan MGB probes contain the following features:
1) A fluorescent reporter at the 5' end
2) A nonfluorescent quencher at the 3' end. Because the quencher does not fluoresce, the real-time instruments can measure the reporter dye contributions more precisely.
3) A minor groove binder at the 3' end. The minor groove binder increases the melting temperature (Tm) of probes, allowing the use of shorter probes.

In general, the TaqMan MGB probes exhibit great differences in Tm values between matched and mismatched probes, which provides more accurate allelic discrimination and makes for a more sensitive real-time assay. Mismatches between a probe and allele, or target, reduce the efficiency of probe hybridization in a measurable way, which is especially important in SNP Genotyping assays. Furthermore, AmpliTaq Gold DNA polymerase is more likely to displace the mismatched probe rather than cleave it to release reporter dye. More information about TaqMan MGB probes can be found in the User Bulletin entitled "Primer Express Version 1.5 and TaqMan MGB Probes for Allelic Discrimination." You can find a copy on our website by entering this title in the main search field.

When using SYBR Green chemistry on an Applied Biosystems Real-Time PCR instrument, how do I change settings to reflect that there is no TaqMan probe being used in the reaction?

Refer to the product manual for your instrument and software for specifics, but in general you will want to change the Quencher value to None.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

How can RNA standards be generated to perform absolute quantitation for RNA targets?

It is generally not possible to use DNA as a standard for absolute quantitation of RNA because there is no control for the efficiency of the reverse transcription step. Therefore, in-vitro transcribed RNA is commonly used to prepare standards for the absolute quantitation of RNA targets. This would involve the cloning of the target of interest into an in-vitro transcription plasmid, performing in-vitro transcription, then purifying the resulting cRNA so that the DNA plasmid cannot serve as a PCR template. Concentration is measured by A260 and converted to the number of copies using the molecular weight of the RNA.

Relative quantitation of gene expression methods require less up-front preparation and provide a fold-change value instead of an absolute quantity result. For many researchers, absolute quantities are not a necessary parameter to measure, and therefore relative quantitation is a much more attractive approach to studying gene expression via real-time PCR. For more information on relative quantitation of gene expression, please refer to our Technical Reference Library in the Technical Resources section of our website.

Citations & References (3)

Citations & References
Abstract
Transcriptional analysis of the human cardiac calsequestrin gene in cardiac and skeletal myocytes.
Authors:Reyes-Juárez JL, Juárez-Rubí R, Rodríguez G, Zarain-Herzberg A,
Journal:J Biol Chem
PubMed ID:17938175
'Calsequestrin is the main calcium-binding protein inside the sarcoplasmic reticulum of striated muscle. In mammals, the cardiac calsequestrin gene (casq2) mainly expresses in cardiac muscle and to a minor extent in slow-twitch skeletal muscle and it is not expressed in non-muscle tissues. This work is the first study on the ... More
Identification and characterization of TriABC-OpmH, a triclosan efflux pump of Pseudomonas aeruginosa requiring two membrane fusion proteins.
Authors:Mima T, Joshi S, Gomez-Escalada M, Schweizer HP,
Journal:J Bacteriol
PubMed ID:17720796
Pseudomonas aeruginosa achieves high-level (MIC>1 mg/ml) triclosan resistance either by constitutive expression of MexAB-OprM, an efflux pump of the resistance nodulation cell division (RND) family, or expression of MexCD-OprJ, MexEF-OprN, and MexJK-OpmH in regulatory mutants. A triclosan-resistant target enzyme and perhaps other mechanisms probably act synergistically with efflux. To probe ... More
Functional analyses of glycyl-tRNA synthetase mutations suggest a key role for tRNA-charging enzymes in peripheral axons.
Authors:Antonellis A, Lee-Lin SQ, Wasterlain A, Leo P, Quezado M, Goldfarb LG, Myung K, Burgess S, Fischbeck KH, Green ED,
Journal:J Neurosci
PubMed ID:17035524
Charcot-Marie-Tooth disease type 2D (CMT2D) and distal spinal muscular atrophy type V (dSMA-V) are axonal neuropathies characterized by a phenotype that is more severe in the upper extremities. We previously implicated mutations in the gene encoding glycyl-tRNA synthetase (GARS) as the cause of CMT2D and dSMA-V. GARS is a member ... More