meso-Erythritol, 99%
meso-Erythritol, 99%
meso-Erythritol, 99%
Thermo Scientific Chemicals

meso-Erythritol, 99%

CAS: 149-32-6 | C4H10O4 | 122.12 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
檢視全部
Plastic Bottle
產品號碼 117820250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
化學識別
CAS149-32-6
IUPAC Namebutane-1,2,3,4-tetrol
Molecular FormulaC4H10O4
InChI KeyUNXHWFMMPAWVPI-UHFFFAOYNA-N
SMILESOCC(O)C(O)CO
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point118°C to 122°C
HPLC>=98.5 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only