Product Image
Invitrogen™
LR Clonase II for BaculoDirect™ Kits
The BaculoDirect™ Baculovirus Expression System is a powerful and versatile eukaryotic system for high-level protein expression in insect cells. The深入閱讀
Have Questions?
產品號碼Quantity
1179102310 reactions
產品號碼 11791023
價格 (TWD)
16,277.00
線上優惠
Ends: 30-Sep-2026
20,346.00
您節省 4,069.00 (20%)
Each
Quantity:
10 reactions
價格 (TWD)
16,277.00
線上優惠
Ends: 30-Sep-2026
20,346.00
您節省 4,069.00 (20%)
Each
The BaculoDirect™ Baculovirus Expression System is a powerful and versatile eukaryotic system for high-level protein expression in insect cells. The combination of Gateway® Technology with baculovirus expression makes the BaculoDirect™ System the fastest and easiest method for production of recombinant baculovirus.
For Research Use Only. Not for use in diagnostic procedures.
規格
Product TypeLR Clonase II
Quantity10 reactions
Shipping ConditionDry Ice
EnzymeLR Clonase
Product LineBaculoDirect, Clonase, Gateway
Unit SizeEach
內容物與存放
Store LR Clonase™ mix at -80°C. All components are guaranteed stable for 6 months if stored properly.
常見問答集 (常見問題)
Can I use E. coli containing an F' plasmid, such as Top 10F', to propagate a vector with the ccdB gene?
While the F' plasmid does contain the ccdA gene that can inhibit or reduce the toxicity of the ccdB gene product, the ccdA expression level is likely to be too low, or inhibition may not be complete, and the bacteria would still be exposed to the ccdB gene product and thus not grow. Therefore, bacterial strains containing the F' plasmid are not recommended as hosts for propagation of ccdB containing vectors.
For propagation of Gateway vectors containing ccdB, we recommend the One Shot ccdB Survival 2 T1R Competent Cells (A10460), which were specifically designed for that purpose. However, please note that these cells are not validated for propagation of other ccdB-containing vectors like the older pZErO plasmids, and in most cases they are not expected to work due to very high levels of ccdB protein expressed in those vectors.