Product Image
Invitrogen™

EXPRESS qPCR Supermix, with premixed ROX

Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’veRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
11795200200 Reactions
1179501K
also known as 11795-01K
1000 Reactions
Catalog number 11795200
Price (USD)
1,064.00
Each
No. of Reactions:
200 Reactions
Price (USD)
1,064.00
Each
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of EXPRESS qPCR SuperMix and added additional capabilities for your gene expression analysis.

EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for your probe-based qPCR assays. This kit comes with premixed ROX reference dye. EXPRESS qPCR SuperMix kits with a separate tube of ROX reference dye are also available.

This kit provides:
• Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
• UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
FormatTube
No. of Reactions200 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymerasePlatinum Taq DNA Polymerase
Product TypeExpress qPCR Supermix
Quantity200 reactions
Reverse TranscriptaseSuperScript™ III
Sample TypeDNA (Genomic), cDNA
Sufficient For200 Reactions
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).