Product Image
Gibco™

CD 293 Medium (1X)

CD 293 Medium is a chemically-defined, protein-free medium optimized for the growth of suspension cultures of 293 cells. CD 293Read more
Have Questions?
Catalog NumberQuantity
119130191000 mL
Catalog number 11913019
Price (ZAR)
2 260.00
Each
Quantity:
1000 mL
Customize this product
Price (ZAR)
2 260.00
Each
CD 293 Medium is a chemically-defined, protein-free medium optimized for the growth of suspension cultures of 293 cells. CD 293 Medium contains no proteins or peptides of animal, plant, or synthetic origin. There are also no undefined lysates or hydrolysates in the formulation.

Formulated without L-glutamine for increased stability; add 20 ml of 200 mM L-glutamine or 20 ml of GlutaMAX™-I Supplement per 1,000 ml of medium prior to use.

A 293 Medium Master file has been submitted to the FDA. If you wish to cross-reference a Master file for CD 293 Medium, please contact Technical Services or your local Sales Representative.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell Line293F, 293 (HEK)
Concentration1 X
Product LineGibco
Product Type293 Medium
Quantity1000 mL
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Low Protein, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
Serum LevelSerum-free
Without AdditivesNo Glutamine
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

How do I adapt my cells to serum-free medium?

Cells can be adapted by Sequential or Direct Adaptation. Suggested protocols for each are below, and you can also find more information by searching "Adaptation of Cell Cultures to a Serum-Free Medium" from our website home page.

SEQUENTIAL ADAPTATION
1) Subculture the cells growing in serum-supplemented medium into a 25%:75% mixture of SFM and serum supplemented medium.
2) When the cell density is 5 x 10E5 cells/ml, subculture the cells into a 50%:50% mixture of SFM and serum supplemented medium at a cell density 2.5 x 10E5 to 3 x 10E5 cells/ml.
3) Continue to subculture after the cell density 5 x 10E5 cells/ml in gradually increasing proportions of SFM until the serum is ~0.1% with about 85% cell viability.
4) Subculture the cells into SFM with an innoculum of 2.5 x 10E5 to 3 x 10E5 cells/ml.
5) When the cell density is 1 x 10E6 to 3 x 10E6 cells/ml (4 to 6 days post planting) subculture the cells again.
6) Stock cultures of SFM adapted cells should be subcultured in SFM every 3 to 5 days when the cell density is 1 x 10E6 to 3 x 10E6 cells/ml with 90% viability.

DIRECT ADAPTATION
Some cells can be directly adapted from serum-containing medium to SFM. For direct adaptation, the cell innoculum should be 1.5 x 10E5 to 3 x 10E5 cells/ml.
Cells should be subcultured when the cell density is 1 x 10E6 to 3 x 10E6 cells/ml. Cells are fully adapted to SFM when the cell density is 2 x 10E6 to 4 x 10E6 cells/ml after 4 to 7 days in culture.
Stock cultures of cells adapted to SFM should be subcultured in SFM every 3 to 5 days when the cell density is 1 x 10E6 to 3 x 10E6 cells/ml with 90% viability.

Why is it necessary to gradually adapt the cells to serum-free medium?

Some cells, such as insect cells, are sensitive to changes in their medium. By sequentially adapting cells, the medium is changed with minimal effects on cell growth.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.