Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD86 Antibodies

          Invitrogen

          CD86 (B7-2) Monoclonal Antibody (GL1), PE, eBioscience™

          98 Published Figures
          218 References
          View all (225) CD86 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD86 (B7-2) Monoclonal Antibody (GL1), PE, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (98)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD86 (B7-2) Antibody (12-0862-81) in Flow

          Staining of 3-day unstimulated (left) and 3 day LPS-stimulated (right) BALB/c splenocytes with 0.06 µg of Rat IgG2a kappa Isotype Control PE (Product # 12-4321-80) (open histogram) or 0.06 µg of Anti-Mouse CD86 (B7-2) PE (filled histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          12-0862-81

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 214 publications 214 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          In vitro Assay (In vitro)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), PE, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          GL1

          Conjugate

          PE PE PE
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 737
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_465767

          Product Specific Information

          Description: The GL1 monoclonal antibody reacts with mouse CD86, an ~80 kDa surface receptor also known as B7-2. CD86 and CD80 are members of the B7 family of costimulatory molecules. CD86 is expressed at low level on B cells, macrophages, and dendritic cells and is upregulated on B cells through a variety of surface stimuli including the BCR complex, CD40 and some cytokine receptors. CD86 is also expressed by activated mouse T cells and thioglycolate-elicited peritoneal cells. In addition to CD80 (B7-1), CD86 is a counter-receptor for the T cell surface molecules CD28 and CD152 (CTLA-4). This interaction plays a critical role in T-B crosstalk, T cell costimulation, autoantibody production and Th2-mediated Ig production. The kinetics of upregulation of CD86 upon stimulation, supports its major contribution during the primary phase of an immune response.

          Applications Reported: The GL1 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This GL1 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD86, along with CD80, is a member of the B7 family of costimulatory molecules and plays a crucial role in T cell activation and immune response regulation. CD86 is expressed at low levels on B cells, macrophages, and dendritic cells, and its expression is upregulated on B cells through various stimuli, including the BCR complex, CD40, and certain cytokine receptors. As a type I membrane protein and member of the immunoglobulin superfamily, CD86 serves as a ligand for the T cell surface proteins CD28 and CTLA-4 (CD152). The interaction between CD86 and CD28 provides a costimulatory signal essential for T cell activation during antigen presentation, while binding with CTLA-4 negatively regulates T cell activation, diminishing the immune response. This interaction is critical for T-B cell crosstalk, T cell costimulation, autoantibody production, and Th2-mediated Ig production. The kinetics of CD86 upregulation upon stimulation suggest its significant contribution during the primary phase of an immune response. CD86 and CD80 have distinct roles in T helper cell differentiation, and insufficient co-stimulation involving these molecules can induce tolerance. Alternative splicing of CD86 results in two transcript variants encoding different isoforms, with additional variants described but not fully sequenced. Dysfunction in CD86 is associated with diseases such as gallbladder squamous cell carcinoma and myocarditis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: activation B7-2 antigen; B lymphocyte activation antigen B72; B7-2; CD28 antigen ligand 2; early T cell costimulatory molecule-1; Early T-cell co-stimulatory molecule 1; MGC34413; unnamed protein product

          View more View less

          Gene Aliases: B7; B7-2; B7.2; B70; Cd28l2; CLS1; ETC-1; Ly-58; Ly58; MB7; MB7-2; TS/A-2

          View more View less

          UniProt ID: (Mouse) P42082

          View more View less

          Entrez Gene ID: (Mouse) 12524

          View more View less

          Function(s)
          receptor binding protein binding receptor agonist activity
          Process(es)
          toll-like receptor signaling pathway positive regulation of immunoglobulin production negative regulation of T cell anergy immune response signal transduction cell surface receptor signaling pathway positive regulation of cell proliferation positive regulation of signal transduction T cell costimulation toll-like receptor 3 signaling pathway positive regulation of T cell proliferation positive regulation of activated T cell proliferation T cell activation B cell activation negative regulation of T cell proliferation lymphocyte activation positive regulation of immune response negative regulation of T cell receptor signaling pathway positive regulation of T cell receptor signaling pathway regulation of T cell activation negative regulation of T cell activation defense response to virus cellular response to lipopolysaccharide positive regulation of NIK/NF-kappaB signaling regulation of leukocyte cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-7d6nq:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline