PureLink™ RNase A (20 mg/mL), 25 mL - FAQs
PureLink™ RNase A (20 mg/mL), 25 mL - FAQs
View additional product information for PureLink™ RNase A (20 mg/mL) - FAQs (12091039, 12091021)
1 product FAQs found
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?
Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.