I am using the PureLink RNA Micro Kit, and my RNA yield is very low. Can you offer some tips?
This could be due to incomplete lysis and homogenization. Here are some recommendations:
- Ensure that 10 µL of 2-mercaptoethanol was added per 1 mL of Lysis Buffer.
- Perform all steps at room temperature unless directed otherwise.
- Decrease the amount of starting material used, or increase volume of Lysis Buffer.
- Use the proper homogenization methods according to recommendations in the sample-specific protocols.
- Cut tissue samples into smaller pieces and ensure the tissue is completely immersed in the Lysis Buffer to achieve optimal lysis.
- Ensure that 10 µL of 2-mercaptoethanol was added per 1 mL of Lysis Buffer.
- Perform all steps at room temperature unless directed otherwise.
- Decrease the amount of starting material used, or increase volume of Lysis Buffer.
- Use the proper homogenization methods according to recommendations in the sample-specific protocols.
- Cut tissue samples into smaller pieces and ensure the tissue is completely immersed in the Lysis Buffer to achieve optimal lysis.