Slit Systems
Imagem ilustrativa. O produto pode variar
Thermo Scientific™
Slit Systems
Enable easy modifications to the width of the ion beam aperture for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system with these slit systems.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoIncludes
1227280Low Resolution Slit Rplcmnt
1170800Plug-in Slit Tongue
1281240Med Resolution Flat Top Peak
Número do catálogo 1227280
Preço (BRL)
-
Includes:
Low Resolution Slit Rplcmnt
Maintain peak performance from your Thermo Scientific™ ELEMENT™ Series HR-ICP-MS analyzer, by using only top quality, dedicated consumables. These high resolution Thermo Scientific™ Slit Systems, defining the ion beam aperture, are manufactured at minimum tolerance specifications, which provides the best user experience for maintaining resolution and reliable interference removal.
Select Quality Consumables for Improved Performance
  • Plug-in Slit Tongue entrance slit – tool-free change of the high-resolution Slit defining Medium and High Mass Resolution
  • Medium Resolution Flat Top Peak Kit; contains a Plug-in Slit tongue for the entrance Slit, as well as a complete exit Slit assembly to achieve flat topped peaks in Medium Resolution. For applications involving Isotope Ratio measurements of Chromium and other transition metals
Especificações
For Use With (Equipment)ELEMENT Series HR-ICP-MS system
IncludesLow Resolution Slit Rplcmnt
TypeLow Resolution Slit Replacement Kit
Unit SizeEach