OptiPRO™ SFM, 100 mL - FAQs
OptiPRO™ SFM, 100 mL - FAQs
View additional product information for OptiPRO™ SFM - FAQs (12309050, 12309019)
4 product FAQs found
Do you offer a protocol for titering virus in OptiPRO SFM?
For virus titering in OptiPRO SFM we recommend using Falcon or equivalent 96-well plates and plating cells at a concentration of 1 X 10^6/mL. If you are having trouble growing cells for virus titration in OptiPRO SFM in the 96-well plates, we suggest switching to the EMEM with 5%FBS or DMEM with 5% FBS.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is there a recommended protocol for culturing cell lines in OptiPRO SFM that require sequential adaptation?
The sequential adaptation process is a multi-subculture of cells. We recommend the following passaging sequence:
• First passage: use 25% OptiPRO SFM:75% original stock medium
• Second passage: use 50% OptiPRO SFM:50% original stock medium
• Third passage: use 75% OptiPRO SFM:25% original stock medium
• Fourth and subsequent passages: use 100% OptiPRO SFM
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I add antibiotics to OptiPRO SFM?
You can supplement OptiPRO SFM with 5mL/L Antibiotic-Antimycotic (Cat. No. 15240096)
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What concentration of L-glutamine or GlutaMAX-I should I add to OptiPRO SFM before I use it?
We recommend adding L-glutamine or GlutaMAX-I to OptiPRO SFM at a final concentration of 4mM.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.