AMV Reverse Transcriptase, cloned - FAQs
AMV Reverse Transcriptase, cloned - FAQs
View additional product information for AMV Reverse Transcriptase, cloned - FAQs (12328019)
1 product FAQs found
How much of the first-strand cDNA reaction should I load for PCR?
While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.