D-Mannitol, 98+%
D-Mannitol, 98+%
D-Mannitol, 98+%
Thermo Scientific Chemicals

D-Mannitol, 98+%

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 125340025
Price (USD)
157.65
Online exclusive
175.00
Save 17.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
157.65
Online exclusive
175.00
Save 17.35
Each
Chemical Identifiers
CAS69-65-8
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Nickel (Ni)=<1 ppm
Appearance (Color)White
Melting point165°C to 170°C
HPLC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only