VP-SFM AGT™ Medium
Gibco™

VP-SFM AGT™ Medium

VP-SFM AGT™ Medium
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
1255900310 kg
125590271 l
1255901910 l
12559001100 L
Katalognummer 12559003
Preis (EUR)
20.300,00
Each
Menge:
10 kg
Customize this product
Preis (EUR)
20.300,00
Each
Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation. VP-SFM is a serum-free, animal-origin-free culture medium designed for growth of VERO cells and virus production. It is also suitable for the growth of COS-7, MDCK, BHK-21, and Hep2 cells and for recombinant protein production.

Features of VP-SFM AGT Medium include:

  • Serum-free, animal-origin-free, ultra-low protein formulation
  • Easy adaptation for many popular cell lines
  • Scalability in flasks, cell factories, and bioreactors with AGT

Serum-free, animal-origin-free, ultra-low protein formulation

VP SFM is formulated with an ultra-low protein concentration (< 5 μg/mL) for easier purification of recombinant proteins.

Easy adaptation for many popular cell lines

VP-SFM was designed for growth of VERO cells.  But it has also been used successfully to grow other cell lines (COS-7, MDCK, BHK-21, and Hep2) and for recombinant protein production, allowing the use of a single medium for multiple cell lines. VP-SFM is formulated without L-glutamine for greater stability and extended shelf life. Simply add L-glutamine or Glutamax supplement as needed.

cGMP manufacturing and quality system

VP-SFM is manufactured in a cGMP-compliant facility. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Scalability with Advanced Granulation Technology

Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration, a just-add-water protocol, and low dust generation, creating less pressure build-up during sterile filtration.

Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
ZelllinieMDCK, VERO, HEp2, BHK-21, COS-7
KulturmilieuCO2
Endotoxin-GehaltLow
FormatRopak Bucket
Glucosekonzentration3.9 g/L
Antibiotika inklusiveKeine Antibiotika
Anorganische SalzeCalcium, Magnesium, Natriumphosphat
ProduktlinieGibco
ProdukttypSerum-freies Medium (SFM)
Reinheits- oder QualitätsgradCertified
Menge10 kg
VersandbedingungDry Ice
KlassifikationChemically-defined, Protein-free, Serum-free
Culture TypeAdherent Cell Culture
FormPulver (AGT)
Serum LevelSerumfrei
SterilitätUnsteril
Mit AdditivenLow Glucose, HEPES, Phenolrot, Sodium Pyruvate, Natriumhydrogencarbonat
Ohne AdditiveKein Glutamin, No Antibiotics
Unit SizeEach
Inhalt und Lagerung
Store at 2 to 8°C in the dark.

Häufig gestellte Fragen (FAQ)

What are the main benefits of using AGT?

Complete formulations
- Homogeneous distribution of minute components
- Complete complex formulations in a dry format, including serum-free, protein-free, and chemically defined nutrient media
- Pre-adjusted to the appropriate pH and osmolality
- Require only standard supplementation with L-glutamine for L-glutamine dependent systems
- Can be customized for modified catalog and customers' proprietary formulations

Granular format
- Dissolves instantly for faster media preparation times
- Dust-free granules reduce mess and environmental contamination
- Performance equivalent to liquid media
- Workflow improvement
- Helps reduce total cycle costs due to lower number of components, decreasing time involved in raw material planning, procurement, and testing
- Faster and easier media preparation, with less potential for error
- Easy to scale up from research through production, in batch sizes from 2 kg to 6,000 kg
- Available in various packaging options

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is AGT?

Granulated cell culture media is an easy-to-use, granular form of dry media that is produced by applying fluid bed granulation technology to cell culture nutrient media manufacturing. The patented process allows production of complete formulations of a variety of serum-free, protein-free, and chemically defined media in a dry format.

AGT offers the benefits of liquid media without the cost, storage, and transportation issues. It is a complete medium that is pH and osmolality pre-adjusted. The granules dissolve instantly for faster media preparation time than conventional dry powder media. AGT media can therefore help reduce total cycle costs, decreasing time involved in raw material planning, procurement, and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.