DL-Methionine, 99+%, Thermo Scientific Chemicals
DL-Methionine, 99+%, Thermo Scientific Chemicals
DL-Methionine, 99+%, Thermo Scientific Chemicals

DL-Methionine, 99+%, Thermo Scientific Chemicals

CAS: 59-51-8 | C5H11NO2S | 149.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic Bottle
Catalog number 125650010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS59-51-8
IUPAC Name2-amino-4-(methylsulfanyl)butanoic acid
Molecular FormulaC5H11NO2S
InChI KeyFFEARJCKVFRZRR-UHFFFAOYNA-N
SMILESCSCCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes
Titration with HClO499.0 to 101.0 % (on dried substance)
Sulfated ash=<0.1 %
Appearance (Color)White to almost white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only