LHC-9 Medium (1X)
LHC-9 Medium (1X)
LHC-9 Medium (1X)
Gibco™

LHC-9 Medium (1X)

LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. For maximum flexibility and convenience,Read more
Have Questions?
Catalog NumberQuantity
12680013500 mL
Catalog number 12680013
Price (USD)
150.65
Online exclusive
162.00
Save 11.35
Each
Quantity:
500 mL
Price (USD)
150.65
Online exclusive
162.00
Save 11.35
Each
LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. For maximum flexibility and convenience, we offer these products with or without supplementation.

LHC Basal Medium (1X), liquid

Applications:
Formulated for the growth of bronchial epithelial cells with further supplements.

Recommended storage conditions:
2° to 8°C.

Shelf life:
12 months

Intended use(s)
Research use only



LHC-8 Medium and LHC-9 Medium


Applications:


Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.


Performance and quality testing:

Performance tested in a growth assay using BEAS-2B cells.

Recommended storage conditions:
-5°C to -20°C.

Intended use(s)
Research use only

Reference(s):
Pawliczak R., et. al. (2001) J Biol Chem 276, 44613. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005. Liu X., et. al. (1998) Am J Physiol 274, L58. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineB3-1, S9, C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), BBE (bovine bronchial epithelial cells)
Cell TypeEpithelial Cells
Concentration1 X
Product TypeLHC-9 Medium
Quantity500 mL
Shelf Life12 Months
Shipping ConditionDry Ice
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.