Luria Broth Base (Miller's LB Broth Base), powder, 500 g - FAQs
Luria Broth Base (Miller's LB Broth Base), powder, 500 g - FAQs
View additional product information for Luria Broth Base (Miller's LB Broth Base), powder - FAQs (12795027, 12795084)
1 product FAQs found
Do you offer an alternative for Luria Broth Base (Miller's LB Broth Base), powder (Cat. No. 12795027, 12795084)?
We do not offer any alternatives for this product.