Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™
Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesFor Use With (Equipment)
1311880Skimmer Cone Insert 2.8 (HS)iCAP Q/Qnova Series ICP-MS
1341430Skimmer Cone Pt Cold/Hot PlasmaiCAP Q/Qnova Series ICP-MS
1341420Skimmer Cone Ni Cold/Hot PlasmaiCAP Q/Qnova Series ICP-MS
1311870Skimmer Cone, Ni, InsertiCAP Q/Qnova Series ICP-MS
1324540Skimmer Cone Insert PtiCAP Q/Qnova Series ICP-MS
1324530Skimmer Cone Insert Pt no BoroniCAP Q/Qnova Series ICP-MS
1318480Skimmer Cone Insert 3.5 (HM)iCAP Q/Qnova Series ICP-MS
BRE0006591Skimmer Cone Insert 4.5 RobustiCAP Q/Qnova Series ICP-MS
3600812Sample Cone, NickeliCAP MX and Q/Qnova ICP-MS
3601289Sample Cone, PlatinumiCAP MX and Q/Qnova ICP-MS
1310900Sample Cone GasketiCAP MX and Q/Qnova ICP-MS
BRE0031487iCAP MX Skimmer Cone NiiCAP MX Series ICP-MS
BRE0063287iCAP MX Skimmer Cone PtiCAP MX Series ICP-MS
BRE0063475iCAP MX Skimmer Cone Pt no BoroniCAP MX Series ICP-MS
Catalog number 1311880
Price (USD)
231.00
Each
Includes:
Skimmer Cone Insert 2.8 (HS)
For Use With (Equipment):
iCAP Q/Qnova Series ICP-MS
Price (USD)
231.00
Each
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
  • Sample cones made from nickel and platinum
  • Skimmer cones made from nickel and platinum
  • Platinum, boron free skimmer cone (for semiconductor-related applications)
  • Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifications
For Use With (Equipment)iCAP Q/Qnova Series ICP-MS
IncludesSkimmer Cone Insert 2.8 (HS)
TypeSkimmer Cone Insert 2,8 (High Sensitivity)
Unit SizeEach