Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution - FAQs
Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution - FAQs
View additional product information for Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution - FAQs (13150016)
1 product FAQs found
What reagents do you offer for cell dissociation, and what are the main differences between them?
Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).
Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.