Injectors for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™

Injectors for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems

Discover our wide range of reliable, screw-in, self-aligning injectors, with differing internal diameters and materials, to match your sample matrices and ICP-MS analysis needs.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoIncluye
1323600Inyector 2,0 mm de D.I., zafiro
1305600Inyector de 2,5 mm de D.I., cuarzo
BRE0017466Inyector de 2,0 mm, cuarzo, 1 entrada
1305640Inyector 2,0 mm de D.I., cuarzo
1321200Inyector 2,0 mm de D.I., platino
1323630Inyector de 2,0 mm, cuarzo, 2 entradas
1325110Inyector 1,0 mm de D.I., cuarzo
1305650Inyector 1,5 mm de D.I., cuarzo
Número de catálogo 1323600
Precio (MXN)
-
Incluye:
Inyector 2,0 mm de D.I., zafiro

Choose from this range of reliable, screw-in, self-aligning injectors, with differing internal diameters and materials, to match your sample matrices and analytical needs.

The iCAP MX and iCAP Q/Qnova Series ICP-MS systems come as standard with a 2.5 mm internal diameter, quartz injector. All of our injectors are easily removable for improved robustness and reduced maintenance requirements.

Choose high-quality consumables to improve performance

     

  • Quartz injectors are suitable for most aqueous acidic and organic applications
  • Sapphire injectors are suitable for highly concentrated acids
  • Platinum injectors are suitable for ultra-trace and highly concentrated acids
  • Injectors with integrated ports are suitable for dry aerosol introduction, e.g., laser ablation

 

Especificaciones
Para utilizar con (equipo)Sistemas ICP-MS iCAP de la serie Q/Qnova
IncluyeInyector 2,0 mm de D.I., zafiro
TipoInyector
Unit SizeEach