Consumables Kits for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™
Consumables Kits for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Consumables Kits include consumables for continual regular maintenance, to ensure your system is achieving maximum performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
1324980iCAP Q/Qnova ICP-MS Systems
BRE0064514iCAP MX Series ICP-MS Systems
Catalog number 1324980
Price (USD)
4,300.00
Each
For Use With (Equipment):
iCAP Q/Qnova ICP-MS Systems
Price (USD)
4,300.00
Each
Use the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Consumables Kits, including consumables for continual regular maintenance, to ensure your system is achieving maximum performance.
Includes the following consumables to run the ICP-MS system for one year:
  • Quartz torch
  • Quartz injector, 2.5 mm
  • Replacement sample and skimmer cones (incl. spare sample cone gaskets)
  • For iCAP Qnova: Replacement inserts (2.8 mm, 3.5 mm, 4.5 mm)
  • Tubing for sample introduction (includes peristaltic pump tubing)
Specifications
DescriptioniCAP Q/Qnova Series Consumables Kit
For Use With (Equipment)iCAP Q/Qnova ICP-MS Systems
IncludesTorch, Inserts, Cones and Sample Introduction Tubing
TypeConsumables Kit
Unit SizeEach