L-Serine methyl ester hydrochloride, 98%
L-Serine methyl ester hydrochloride, 98%
L-Serine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Serine methyl ester hydrochloride, 98%

CAS: 5680-80-8 | C4H10ClNO3 | 155.578 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 132670050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS5680-80-8
IUPAC Namemethyl 2-amino-3-hydroxypropanoate hydrochloridyl
Molecular FormulaC4H9ClNO3
InChI KeyQDXGLJWDLGMECL-UHFFFAOYNA-N
SMILES[Cl].COC(=O)C(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Argentometric>=97.5 %
Sulfated ash=<0.1 %
Water=<1.0 % (K.F.)
Appearance of solution(5% in methanol) clear colorless to almost colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only