This product ships via ground from US northeast.
D-Sorbitol, 97%
D-Sorbitol, 97%
D-Sorbitol, 97%
Thermo Scientific Chemicals

D-Sorbitol, 97%

CAS: 50-70-4 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 132735000
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.30
Save 5.65
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.30
Save 5.65
Each
Chemical Identifiers
CAS50-70-4
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Sulfated ash=<0.1 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Specific optical rotation(on anhydrous substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only