Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Actual product may vary
Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™
Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesFor Use With (Equipment)
1341430Skimmer Cone Pt Cold/Hot PlasmaiCAP Q/Qnova Series ICP-MS
1341420Skimmer Cone Ni Cold/Hot PlasmaiCAP Q/Qnova Series ICP-MS
1311870Skimmer Cone, Ni, InsertiCAP Q/Qnova Series ICP-MS
1324540Skimmer Cone Insert PtiCAP Q/Qnova Series ICP-MS
1324530Skimmer Cone Insert Pt no BoroniCAP Q/Qnova Series ICP-MS
1311880Skimmer Cone Insert 2.8 (HS)iCAP Q/Qnova Series ICP-MS
1318480Skimmer Cone Insert 3.5 (HM)iCAP Q/Qnova Series ICP-MS
BRE0006591Skimmer Cone Insert 4.5 RobustiCAP Q/Qnova Series ICP-MS
3600812Sample Cone, NickeliCAP MX and Q/Qnova ICP-MS
3601289Sample Cone, PlatinumiCAP MX and Q/Qnova ICP-MS
1310900Sample Cone GasketiCAP MX and Q/Qnova ICP-MS
BRE0031487iCAP MX Skimmer Cone NiiCAP MX Series ICP-MS
BRE0063287iCAP MX Skimmer Cone PtiCAP MX Series ICP-MS
BRE0063475iCAP MX Skimmer Cone Pt no BoroniCAP MX Series ICP-MS
Catalog number 1341430
Price (USD)
3,280.00
Each
Includes:
Skimmer Cone Pt Cold/Hot Plasma
For Use With (Equipment):
iCAP Q/Qnova Series ICP-MS
Price (USD)
3,280.00
Each
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
  • Sample cones made from nickel and platinum
  • Skimmer cones made from nickel and platinum
  • Platinum, boron free skimmer cone (for semiconductor-related applications)
  • Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifications
For Use With (Equipment)iCAP Q/Qnova Series ICP-MS
IncludesSkimmer Cone Pt Cold/Hot Plasma
TypeSkimmer Cone
Unit SizeEach