Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Tau Antibodies

          Invitrogen

          Tau Monoclonal Antibody (T46)

          Advanced Verification
          33 Published Figures
          60 References
          View all (227) Tau antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Tau Monoclonal Antibody (T46)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (33)
          • Advanced Verification (2)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 42

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Tau Antibody (13-6400) in ICC/IF

          Immunofluorescent analysis of Tau was done on 70% confluent log phase SHSY5Y cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with Tau Mouse Monoclonal Antibody (Product # 13-6400) at 1:250 dilution in 1% BSA and incubated for 3 hours at room temperature and then labeled with Alexa Fluor 488 Rabbit Anti-Mouse IgG Secondary Antibody (Product # A-11059) at a dilution of 1:400 for 30 minutes... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Tau Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Tau Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Tau Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Immunohistochemistry (IHC)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunocytochemistry (ICC/IF)
          Tau Antibody in Immunohistochemistry (IHC)
          Tau Antibody in Immunohistochemistry (IHC)
          Tau Antibody in Immunohistochemistry (IHC)
          Tau Antibody in Immunohistochemistry (IHC)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody in Western Blot (WB)
          Tau Antibody
          Tau Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          13-6400

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-1.0 µg/mL
          View 32 publications 32 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:250
          View 14 publications 14 publications

          ELISA (ELISA)

          0.1-0.5 µg/mL
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          2-5 µg
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Dot blot (DB)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Human, Mouse, Rabbit, Rat, Xenopus

          Published species

          Fruit fly, Human, Mouse, Non-human primate, Rat, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          T46

          Immunogen

          Bovine Tau.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533025

          Product Specific Information

          This antibody recognizes an epitope near the C-terminal region (404-441 aa) of human tau. It reacts with all 7 isoforms of human tau (binding-competent and binding-incompetent), A68 (an abnormal phosphorylated derivative of tau), bovine tau, and human NeuroFibrillary Tangles (NFTs).

          Reactivity is independent of the state of tau phosphorylation. Some cross-reactivity with MAP2 is also observed.

          This antibody is suitable for immunohistochemical staining of Bouin's-fixed or alcohol-fixed, paraffin-embedded or frozen tissue sections.

          Target Information

          Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FLJ31424; FTDP17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; MAPT; MAPT_BigTau; MGC138549; microtubule-associated protein tau; microtubule-associated protein tau 0N4R; microtubule-associated protein tau 1N4R; microtubule-associated protein tau 2N4R; microtubule-associated protein tau BigTau; microtubules; neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHF-tau; PHFtau; protein phosphatase 1, regulatory subunit 103; tau microtubule binding protein; Tau microtubule-associated protein; Tau-4; Tau-derived paired helical filament hexapeptide; Tau5; unnamed protein product

          View more View less

          Gene Aliases: DDPAC; FTD1; FTDP-17; MAPT_0N4R; MAPT_1N4R; MAPT_2N4R; MAPTL; MSTD; Mtapt; MTBT1; MTBT2; PHF-tau; PPND; PPP1R103; pTau; RNPTAU; TAU; tau-40; Tau-PHF6

          View more View less

          UniProt ID: (Bovine) P29172, (Human) P10636, (Mouse) P10637

          View more View less

          Entrez Gene ID: (Bovine) 281296, (Chicken) 426737, (Rabbit) 100344003, (Human) 4137, (Mouse) 17762, (Rat) 29477

          View more View less

          Function(s)
          protein binding DNA binding AT DNA binding double-stranded DNA binding single-stranded DNA binding RNA binding actin binding enzyme inhibitor activity microtubule binding tubulin binding SH3 domain binding enzyme binding protein kinase binding protein binding, bridging apolipoprotein binding dynactin binding phosphatidylinositol binding identical protein binding sequence-specific DNA binding chaperone binding protein phosphatase 2A binding Hsp90 protein binding lipoprotein particle binding histone-dependent DNA binding microtubule lateral binding microtubule stabilizing activity phosphatidylinositol bisphosphate binding heat shock protein binding macromolecular complex binding
          Process(es)
          positive regulation of diacylglycerol kinase activity microtubule cytoskeleton organization microglial cell activation cellular response to DNA damage stimulus cell-cell signaling synapse assembly learning or memory memory response to lead ion regulation of autophagy negative regulation of gene expression negative regulation of mitochondrial membrane potential rRNA metabolic process axonal transport of mitochondrion central nervous system neuron development regulation of microtubule polymerization or depolymerization regulation of microtubule polymerization positive regulation of microtubule polymerization cytoplasmic microtubule organization neuron projection development positive regulation of superoxide anion generation regulation of chromosome organization stress granule assembly cellular response to heat cellular response to reactive oxygen species positive regulation of axon extension microtubule polymerization astrocyte activation regulation of synaptic plasticity intracellular distribution of mitochondria generation of neurons synapse organization regulation of calcium-mediated signaling protein polymerization protein homooligomerization regulation of microtubule-based movement axon development regulation of microtubule cytoskeleton organization plus-end-directed organelle transport along microtubule regulation of mitochondrial fission negative regulation of mitochondrial fission fibril organization axonal transport regulation of cellular response to heat regulation of long term synaptic depression positive regulation of protein localization to synapse neurofibrillary tangle assembly negative regulation of establishment of protein localization to mitochondrion positive regulation of cellular protein localization amyloid fibril formation cellular response to nerve growth factor stimulus cellular response to brain-derived neurotrophic factor stimulus neuron migration axonogenesis response to nutrient adult walking behavior axo-dendritic transport intrinsic apoptotic signaling pathway in response to oxidative stress positive regulation of mitochondrial membrane potential positive regulation of neuron projection development negative regulation of intracellular transport mitochondrion transport along microtubule axon extension mRNA transport response to hyperoxia positive regulation of long term synaptic depression negative regulation of tubulin deacetylation positive regulation of mesenchymal stem cell migration microtubule-based process female pregnancy response to gamma radiation neuron differentiation response to ethanol response to arsenic-containing substance cellular response to calcium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch