Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Transferrin Receptor (CD71) Antibodies

          Invitrogen

          Transferrin Receptor Monoclonal Antibody (H68.4)

          Advanced Verification
          92 Published Figures
          820 References
          View all (178) Transferrin Receptor (CD71) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Transferrin Receptor Monoclonal Antibody (H68.4)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (92)
          • Advanced Verification (2)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 98

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Transferrin Receptor Antibody (13-6890) in ICC/IF

          Immunofluorescence analysis of Transferrin Receptor Monoclonal Antibody (H68.4) was performed using 70% confluent log phase MCF-7 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Transferrin Receptor (H68.4) Mouse Monoclonal Antibody (Product # 13-6800... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Transferrin Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Transferrin Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Transferrin Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Immunocytochemistry (ICC/IF)
          Transferrin Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Transferrin Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Transferrin Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Transferrin Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Transferrin Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Transferrin Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Immunohistochemistry (IHC)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody in Western Blot (WB)
          Transferrin Receptor Antibody
          Transferrin Receptor Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          13-6890

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5 µg/mL
          View 485 publications 485 publications

          Immunohistochemistry (IHC)

          -
          View 38 publications 38 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          View 11 publications 11 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:250
          View 151 publications 151 publications

          Flow Cytometry (Flow)

          -
          View 17 publications 17 publications

          ELISA (ELISA)

          Assay-dependent
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 19 publications 19 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Control (Ctrl)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          Assay-dependent
          View 3 publications 3 publications

          In vitro Assay (In vitro)

          -
          View 1 publication 1 publication

          GST Pull Down (GST-PD)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 87 publications 87 publications
          Product Specifications

          Species Reactivity

          Chicken, Hamster, Human, Mouse, Rat

          Published species

          Bovine, C. elegans, Cat, Chicken, Dog, Hamster, Horse, Human, Insect, Mammal, Mouse, Non-human primate, Pig, Rat, Virus, Yeast, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          H68.4

          Immunogen

          Recombinant human transferrin receptor.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          500 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533030

          Product Specific Information

          H68.4 is specific to residues 3-28 of the human transferrin receptor (TfR) tail.

          Target Information

          Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD antigen CD71; mammary tumor virus receptor 1; tfR; tfR1; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 1; transferrin receptor 2; trfr; unnamed protein product

          View more View less

          Gene Aliases: 2610028K12Rik; CD71; E430033M20Rik; I79_000642; IMD46; Mtvr1; p90; T9; TFR; TFR1; TFR2; TR; TRFR

          View more View less

          UniProt ID: (Human) P02786, (Chicken) Q90997, (Rat) Q99376, (Mouse) Q62351, (Chinese hamster) Q07891

          View more View less

          Entrez Gene ID: (Human) 7037, (Chicken) 396191, (Rat) 64678, (Mouse) 22042, (Chinese hamster) 100689395

          View more View less

          Function(s)
          RNA binding double-stranded RNA binding transferrin receptor activity protein binding protein kinase binding Hsp70 protein binding identical protein binding protein homodimerization activity macromolecular complex binding chaperone binding iron ion transmembrane transporter activity metalloprotease
          Process(es)
          regulation of cell growth response to hypoxia iron ion transport cellular iron ion homeostasis receptor-mediated endocytosis acute-phase response cell surface receptor signaling pathway response to nutrient response to iron ion response to manganese ion positive regulation of gene expression negative regulation of mitochondrial fusion osteoclast differentiation positive regulation of B cell proliferation positive regulation of protein complex assembly receptor internalization response to retinoic acid transferrin transport intracellular signal transduction positive regulation of T cell proliferation regulation of cell proliferation negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of isotype switching response to copper ion multicellular organismal iron ion homeostasis cellular response to xenobiotic stimulus regulation of postsynaptic membrane neurotransmitter receptor levels transport across blood-brain barrier positive regulation of protein localization to nucleus cellular response to leukemia inhibitory factor iron ion transmembrane transport cellular response to iron ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch