L-Threonine, 98%
L-Threonine, 98%
L-Threonine, 98%
Thermo Scientific Chemicals

L-Threonine, 98%

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Catalog number 138930050
Price (USD)
24.60
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
24.60
Each
Chemical Identifiers
CAS72-19-5
IUPAC Name2-amino-3-hydroxybutanoic acid
Molecular FormulaC4H9NO3
InChI KeyAYFVYJQAPQTCCC-UHFFFAOYNA-N
SMILESCC(O)C(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or glistening flakes
Titration with HClO497.5 to 102.5 % (On dried substance)
Heavy metals (as Pb)=<10 ppm
Appearance (Color)White to off-white
Loss on drying=<0.5 % (1 g, 105°C, 3 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only