Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • alpha Catenin Antibodies

          Invitrogen

          alpha Catenin Monoclonal Antibody (alpha-CAT-7A4)

          Advanced Verification
          13 Published Figures
          44 References
          View all (11) alpha Catenin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite alpha Catenin Monoclonal Antibody (alpha-CAT-7A4)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          • Published Figures (13)
          • Advanced Verification (4)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          alpha Catenin Antibody (13-9700) in ICC/IF

          Immunofluorescent analysis of alpha Catenin was performed using 90% confluent log phase MCF-7 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with alpha Catenin (alpha-CAT-7A4) Mouse Monoclonal Antibody (Product # 13-9700) at 1:250 dilution in0.1% BSA and ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Immunohistochemistry (IHC)
          alpha Catenin Antibody in Immunohistochemistry (IHC)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunohistochemistry (IHC)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody
          alpha Catenin Antibody
          alpha Catenin Antibody
          alpha Catenin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          alpha Catenin Monoclonal Antibody (alpha-CAT-7A4)

          Product Details

          13-9700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:250
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 4 publications 4 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:250
          View 16 publications 16 publications

          Flow Cytometry (Flow)

          3-5 µg/1x10^6 cells
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Rat, Xenopus

          Published species

          Bovine, Dog, Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          alpha-CAT-7A4

          Immunogen

          Synthetic peptide derived from the C-terminus of mouse alpha-catenin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533044

          Target Information

          Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 102 kDa cadherin-associated protein; alpha E-catenin; alpha N-catenin; alpha(N)-catenin; alpha-catenin; alpha-catenin-related protein; alpha-E-catenin; alpha-N-catenin; alphaE-catenin; alphaN-catenin; cadherin associated protein; cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha-1-like; Catenin alpha1; Catenin alpha2; Catenin-alpha1; Catenin-alpha2; cerebellar deficient folia; epididymis secretory sperm binding protein; FLJ36832; FLJ52416; HGNC:2509; neural alpha-catenin; NY-REN-13 antigen; renal carcinoma antigen NY-REN-13; unnamed protein product

          View more View less

          Gene Aliases: 2010010M04Rik; CAP-R; CAP102; CAPR; Catna; Catna1; Catna2; CDCBM9; cdf; chp; CT114; CTNR; MDBS2; MDPT2

          View more View less

          UniProt ID: (Human) P35221, (Chicken) P30997, (Human) P26232, (Mouse) P26231, (Mouse) Q61301

          View more View less

          Entrez Gene ID: (Chicken) 416184, (Human) 1495, (Chicken) 396035, (Human) 1496, (Rat) 307505, (Mouse) 12385, (Mouse) 12386, (Rat) 297357

          View more View less

          Function(s)
          RNA binding structural molecule activity protein binding beta-catenin binding cytoskeletal protein binding vinculin binding identical protein binding macromolecular complex binding gamma-catenin binding cadherin binding actin filament binding structural constituent of cytoskeleton non-motor actin binding protein
          Process(es)
          ovarian follicle development cell adhesion male gonad development gap junction assembly cell migration axon regeneration odontogenesis of dentin-containing tooth apical junction assembly response to estrogen cellular response to indole-3-methanol cell-cell adhesion negative regulation of protein localization to nucleus neural crest cell migration axonogenesis protein localization epithelial cell differentiation dendrite morphogenesis brain morphogenesis regulation of synapse structural plasticity trigeminal ganglion formation cytoskeleton organization regulation of neuron projection development radial glia guided migration of Purkinje cell negative regulation of Arp2/3 complex-mediated actin nucleation prepulse inhibition modification of postsynaptic actin cytoskeleton regulation of neuron migration establishment or maintenance of cell polarity negative regulation of neuroblast proliferation regulation of cell proliferation negative regulation of apoptotic process positive regulation of smoothened signaling pathway epithelial cell-cell adhesion negative regulation of cell motility negative regulation of integrin-mediated signaling pathway negative regulation of extrinsic apoptotic signaling pathway in absence of ligand positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch