Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD80 (B7-1) Antibodies

          Invitrogen

          CD80 (B7-1) Monoclonal Antibody (16-10A1), eBioscience™

          72 Published Figures
          99 References
          View all (172) CD80 (B7-1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD80 (B7-1) Monoclonal Antibody (16-10A1), eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (72)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 73

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD80 (B7-1) Antibody (14-0801-81) in Flow

          Staining of 3-day LPS-stimulated BALB/c splenocytes with 0.25 µg of Armenian Hamster IgG Isotype Control Purified (Product # 14-4888-81) (open histogram) or 0.25 µg of Anti-Mouse CD80 (B7-1) Purified (filled histogram) followed by Anti-Armenian Hamster IgG FITC (Product # 11-4111). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Immunocytochemistry (ICC/IF)
          CD80 (B7-1) Antibody in Immunocytochemistry (ICC/IF)
          CD80 (B7-1) Antibody in Immunohistochemistry (IHC)
          CD80 (B7-1) Antibody in Immunohistochemistry (IHC)
          CD80 (B7-1) Antibody in Immunohistochemistry (IHC)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)
          CD80 (B7-1) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-0801-81

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.5 µg/test
          View 92 publications 92 publications

          Immunoprecipitation (IP)

          Assay-Dependent
          -

          Neutralization (Neu)

          Assay-Dependent
          View 1 publication 1 publication

          Functional Assay (Functional)

          Assay-Dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Mouse, Pig

          Published species

          Chicken, Mouse, Pig

          Host/Isotype

          Armenian hamster / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          16-10A1

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • Brilliant UV 615
          • Brilliant UV 737
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467345

          Product Specific Information

          Description: The 16-10A1 monoclonal antibody reacts with mouse CD80 (B7-1), a 55 kDa member of the Ig superfamily. CD80 is expressed by macrophages, dendritic cells and activated B cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively.

          Applications Reported: This 16-10A1 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of frozen tissue sections. 16-10A1 has also been reported in blocking of CD80 to its ligands. (Please use Functional Grade purified 16-10A1, Product # 16-0801, in functional assays).

          Applications Tested: The 16-10A1 antibody has been tested by flow cytometric analysis of activated mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: activation B7-1 antigen; B7 protein; B7-1 protein; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); sCD 80; sCD80; soluble CD 80; soluble CD80; T-cell co-stimulatory protein B7-1; T-cell costimulatory molecule; TS/A-1; unnamed protein product

          View more View less

          Gene Aliases: B7-1; B71; Cd28l; Ly-53; Ly53; MIC17; TSA1

          View more View less

          UniProt ID: (Mouse) Q00609

          View more View less

          Entrez Gene ID: (Dog) 403765, (Pig) 397161, (Mouse) 12519

          View more View less

          Function(s)
          protein binding coreceptor activity receptor agonist activity immunoglobulin receptor superfamily
          Process(es)
          immune system process positive regulation of immune system process negative regulation of T cell mediated immunity immune response cell surface receptor signaling pathway T cell costimulation positive regulation of T cell proliferation T cell activation negative regulation of T cell proliferation positive regulation of alpha-beta T cell proliferation positive regulation of peptidyl-tyrosine phosphorylation negative regulation of T cell receptor signaling pathway positive regulation of T cell receptor signaling pathway regulation of T cell activation negative regulation of T cell activation cellular response to lipopolysaccharide regulation of leukocyte cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0