Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NG2 Antibodies

          Invitrogen

          Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (9.2.27), eBioscience™

          Advanced Verification
          6 Published Figures
          5 References
          View all (43) NG2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (9.2.27), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (6)
          • Advanced Verification (2)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Neural/Glial Antigen 2 (NG2) Antibody (14-6504-37) in ICC/IF

          Immunofluorescence analysis of NG2 was performed using 70% confluent log phase A-375 cells. The cells were fixed with 4% Paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 10 minutes at room temperature. The cells were labeled with Anti-NG2 Monoclonal Antibody (Product # 14-6504-80) at 10 µg/mL in 0.1% BSA, incubated at 4 degree Celsius overnig... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Neural/Glial Antigen 2 (NG2) Antibody in Western Blot (WB)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunocytochemistry (ICC/IF)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunohistochemistry (IHC)
          Neural/Glial Antigen 2 (NG2) Antibody in Immunohistochemistry (IHC)
          Neural/Glial Antigen 2 (NG2) Antibody
          Neural/Glial Antigen 2 (NG2) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-6504-37

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          View 3 publications 3 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          -

          Immunoprecipitation (IP)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Rat

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          9.2.27

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          2 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2865203

          Product Specific Information

          Description: This 9.2.27 monoclonal antibody reacts with human neural/glial antigen 2, which is also known as melanoma chondroitin sulfate proteoglycan (MCSP). This antigen is composed of a 250 kDa N-linked glycoprotein and a >450 kDa proteoglycan. NG2 is present on the surface of >90% of malignant melanomas in addition to some non-melanomic tumors. Moreover, NG2 is expressed on glioma cells, as well as on developing and adult oligodendrocyte precursor cells. Studies have demonstrated the involvement of this protein in tumor cell proliferation, adhesion, migration, and invasion. NG2 also functions as a coreceptor for alpha4beta1 integrin. Finally, expression of NG2 can be used as a prognostic marker for disorders such as acral letiginous melanoma and infantile acute myeloid leukemia.

          The 9.2.27 antibody has been reported to suppress melanoma tumor growth.

          Applications Reported: This 9.2.27 has been reported for use in flow cytometric analysis, immunoprecipitation, immunohistochemical staining of formalin-fixed paraffin embedded tissue (IHC-P), and immunocytochemical staining of fixed cells (ICC).

          Applications Tested: This 9.2.27 antibody has been tested by immunocytochemistry on methanol-fixed A375 cells at less than or equal to 10 µg/mL and by flow cytometric analysis on A375 cells at less than or equal to 0.125 µg/test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10e5 to 10e8 cells/test. It is recommended that this antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cell surface proteoglycan; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; HMW MAA; MEL CSPG; melanoma chondroitin sulfate proteoglycan; melanoma-associated chondroitin sulfate proteoglycan

          View more View less

          Gene Aliases: CSPG4A; HMW-MAA; MCSP; MCSPG; MEL-CSPG; MSK16; NG2

          View more View less

          UniProt ID: (Human) Q6UVK1

          View more View less

          Entrez Gene ID: (Human) 1464

          View more View less

          Function(s)
          protein binding coreceptor activity protein kinase binding
          Process(es)
          signal transduction nervous system development intracellular signal transduction positive regulation of peptidyl-tyrosine phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-fr8qn:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0