Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • Nidogen Proteins

          Gibco

          Human Nidogen Recombinant Protein, PeproTech®

          View all (2) Nidogen proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human Nidogen Recombinant Protein, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          140-17-5UG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          ELISA standard (ELISA Std)

          Assay-dependent
          -

          Binding assay (Binding)

          Assay-dependent
          -
          Product Specifications

          Species

          Human

          Expression System

          CHO cells

          Amino acid sequence

          LSRQELFPFG PGQGDLELED GDDFVSPALE LSGALRFYDR SDIDAVYVTT NGIIATSEPP AKESHPGLFP PTFGAVAPFL ADLDTTDGLG KVYYREDLSP SITQRAAECV HRGFPEISFQ PSSAVVVTWE SVAPYQGPSR DPDQKGKRNT FQAVLASSDS SSYAIFLYPE DGLQFHTTFS KKENNQVPAV VAFSQGSVGF LWKSNGAYNI FANDRESVEN LAKSSNSGQQ GVWVFEIGSP ATTNGVVPAD VILGTEDGAE YDDEDEDYDL ATTRLGLEDV GTTPFSYKAL RRGGADTYSV PSVLSPRRAA TERPLGPPTE RTRSFQLAVE TFHQQHPQVI DVDEVEETGV VFSYNTDSRQ TCANNRHQCS VHAECRDYAT GFCCSCVAGY TGNGRQCVAE GSPQRVNGKV KGRIFVGSSQ VPIVFENTDL HSYVVMNHGR SYTAISTIPE TVGYSLLPLA PVGGIIGWMF AVEQDGFKNG FSITGGEFTR QAEVTFVGHP GNLVIKQRFS GIDEHGHLTI DTELEGRVPQ IPFGSSVHIE PYTELYHYST SVITSSSTRE YTVTEPERDG ASPSRIYTYQ WRQTITFQEC VHDDSRPALP STQQLSVDSV FVLYNQEEKI LRYALSNSIG PVREGSPDAL QNPCYIGTHG CDTNAACRPG PRTQFTCECS IGFRGDGRTC YDIDECSEQP SVCGSHTICN NHPGTFRCEC VEGYQFSDEG TCVAVVDQRP INYCETGLHN CDIPQRAQCI YTGGSSYTCS CLPGFSGDGQ ACQDVDECQP SRCHPDAFCY NTPGSFTCQC KPGYQGDGFR CVPGEVEKTR CQHEREHILG AAGATDPQRP IPPGLFVPEC DAHGHYAPTQ CHGSTGYCWC VDRDGREVEG TRTRPGMTPP CLSTVAPPIH QGPAVPTAVI PLPPGTHLLF AQTGKIERLP LEGNTMRKTE AKAFLHVPAK VIIGLAFDCV DKMVYWTDIT EPSIGRASLH GGEPTTIIRQ DLGSPEGIAV DHLGRNIFWT DSNLDRIEVA KLDGTQRRVL FETDLVNPRG IVTDSVRGNL YWTDWNRDNP KIETSYMDGT NRRILVQDDL GLPNGLTFDA FSSQLCWVDA GTNRAECLNP SQPSRRKALE GLQYPFAVTS YGKNLYFTDW KMNSVVALDL AISKETDAFQ PHKQTRLYGI TTALSQCPQG HNYCSVNNGG CTHLCLATPG SRTCRCPDNT LGVDCIEQK

          Molecular weight

          133.5 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 95% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by the ability of the immobilized protein to support the adhesion of HUVEC cells.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          5 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Target Information

          This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: enactin; entactin; epididymis secretory sperm binding protein; NID 1; NID-1; Nidogen (enactin); Nidogen-1;; unnamed protein product

          View more View less

          Gene Aliases: NID

          View more View less

          UniProt ID: (Human) P14543

          View more View less

          Entrez Gene ID: (Human) 4811

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0