HBSS, calcium, magnesium, no phenol red
Gibco™

HBSS, calcium, magnesium, no phenol red

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
140250761000 mL
14025092500 mL
1402513410 x 500 mL
140251266 x 1000 mL
Catalog number 14025076
Price (USD)
51.65
Online exclusive
55.00
Save 3.35
Each
Quantity:
1000 mL
Customize this product
Price (USD)
51.65
Online exclusive
55.00
Save 3.35
Each

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Calcium, Magnesium, Glucose

Without: Phenol Red, Sodium Pyruvate

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialHanks' Balanced Salt Solution (HBSS)
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 to 305 mOsm/kg
Recommended StorageStorage conditions: 15–30°C, Shipping conditions: Ambient, Shelf life: 36 months from date of manufacture
SterilitySterile
Sterilization MethodSterile-filtered
Concentration1X
For Use With (Application)Mammalian Cell Culture
GradeCell Culture
Physical FormLiquid
Product LineGibco
Quantity1000 mL
Solution TypepH Buffer, Storage Solution and Cleaning Solution
pH6.7 to 7.8
Unit SizeEach

Frequently asked questions (FAQs)

What is the pH of HBSS, calcium, magnesium, no phenol red at room temperature.

At room temperature, HBSS, calcium, magnesium, no phenol red has a pH of 6.7 - 7.8.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?

The recommended storage condition for our HBSS products is at 15-30 degrees C. Generally, storage at 4 degrees C will not impact these products. The only concern would be for the 10x HBSS solutions with calcium and magnesium as these salts might precipitate when stored outside the recommended temperature ranges. As long as you don't see any precipitation upon refrigeration, the product will be fine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I need to add additional HEPES to Hank's Balanced Salt Solution (HBSS) (Cat. No. 14025xxx)?

HBSS (Cat. No. 14025xxx) does not contain HEPES and it needs to be externally supplemented. The complete formulation is available here (https://www.thermofisher.com/us/en/home/technical-resources/media-formulation.153.html).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Why do you offer HBSS with and without phenol red?

Phenol red is a pH indicator which allows you to determine the optimal atmospheric conditions for your cell line during your particular assays. We offer both versions to allow you the option to utilize this indicator in your media.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I use HBSS (with calcium and magnesium) to wash my cells before dissociation?

No, a saline solution that contains calcium and magnesium should not be used to wash cells before dissociation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Multicolor and electron microscopic imaging of connexin trafficking.
Authors: Gaietta Guido; Deerinck Thomas J; Adams Stephen R; Bouwer James; Tour Oded; Laird Dale W; Sosinsky Gina E; Tsien Roger Y; Ellisman Mark H;
Journal:Science
PubMed ID:11964472
'Recombinant proteins containing tetracysteine tags can be successively labeled in living cells with different colors of biarsenical fluorophores so that older and younger protein molecules can be sharply distinguished by both fluorescence and electron microscopy. Here we used this approach to show that newly synthesized connexin43 was transported predominantly in ... More