L(-)-Tryptophan, 99%
L(-)-Tryptophan, 99%
L(-)-Tryptophan, 99%
Thermo Scientific Chemicals

L(-)-Tryptophan, 99%

L(-)-Tryptophan, CAS # 73-22-3, is an essential amino acid necessary for the biosynthesis of proteins. | CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
1 kg
Packaging:
View all
Plastic Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 140590010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS73-22-3
IUPAC Name(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular FormulaC11H12N2O2
InChI KeyQIVBCDIJIAJPQS-VIFPVBQESA-N
SMILESN[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation-29.0° to -33.0° (20°C, 589 nm) (C=1, H2O)
Appearance (Form)Crystalline powder
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Color)White to yellowish-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L(-)-Tryptophan is a nonpolar aromatic compound and essential amino acid required for protein synthesis
  • It is a biochemical precursor of niacin, indole alkaloids, and serotonin

Applications

  • L-Tryptophan acts as a natural dietary supplement and has laboratory applications in the research of antidepressants, anxiolytics, and sleep aids
  • It is a precursor of metabolites that are important in the laboratory study of mammalian physiology, including gastrointestinal functions, immunity, metabolism, and the nervous system
RUO – Research Use Only