Pierce™ 96-Well Polystyrene Plates, White Opaque - FAQs
Pierce™ 96-Well Polystyrene Plates, White Opaque - FAQs
View additional product information for Pierce™ 96-Well Polystyrene Plates, White Opaque - FAQs (15042)
1 product FAQs found
What kind of 96-well plate do I need for my fluorescent assay?
Select a microplate that is compatible with your instrument. Refer to the manufacturer's manual for types of microplates that are recommended for use with your instrument.
Various types of plastic may autofluoresce. Select a microplate that is rated for fluorescence-based assays and exhibits little or no autofluorescence in the emission range of the reagent you wish to use.
Clear plates are routinely used for fluorescence-based assays, but opaque-walled microplates reduce bleed-through between wells. If you are using live cells, you may need to consider special substrate treatments for the bottom of the wells, such as poly-L-lysine.