Pierce™ Copper Coated High Capacity Plates, Black, 96-Well, 5 plates - FAQs
Pierce™ Copper Coated High Capacity Plates, Black, 96-Well, 5 plates - FAQs
View additional product information for Pierce™ Copper Coated High Capacity Plates - FAQs (15146, 15143, 15148, 15147)
1 product FAQs found
Do Pierce Copper Coated High Capacity Plates, White, 96-Well (Cat. No. 15147) and Pierce Copper Coated High Capacity Plates, Black, 96-Well (Cat. No. 15148) have clear bottoms to be read from below?
Pierce Copper Coated High Capacity Plates, White, 96-Well (Cat. No. 15147) and Pierce Copper Coated High Capacity Plates, Black, 96-Well (Cat. No. 15148) can only be read from the top as they have opaque bottoms.