What is the detection limit of proteins with Pierce Glutathione Coated Plates?
The background is extremely low when using the Pierce Glutathione Coated Plates. This results in a high signal-to-noise ratio and a low detection limit which in general means that the detection limit (OD 0.5) is 3 ng per well (100 µL).
How long are Pierce Glutathione Coated Plates stable?
The Pierce Glutathione Coated Plates are extremely stable over long periods of time, if stored at room temperature (20-25 degrees C). We guarantee 12 mths of shelf life in unopened foil package after the date of shipment.
Can I determine the concentration of an unknown GST-tagged protein?
To determine the approximate concentration of a GST-tagged protein, a standard curve should be generated using purified GST added in different concentrations to the wells of the plates. Please note that it is only the approximately concentration that can be determined since not 100% of the GST-fusion proteins will be bound due to denaturing or steric hindrance to GSH.
Can I whirl-mix solutions containing GST proteins?
No. Do not whirl-mix a solution containing GST-tagged proteins. The CV will increase due to slight degradation of the GST protein.
With Pierce Glutathione Coated Plates, why should I use Tween-20 surfactant in the washing step and in the buffers containing the antibodies?
We recommend using 0.05% Tween-20 surfactant (as the non-ionic detergent) to improve the signal-to-noise ratio of the assay and thus decrease the background.
Should I optimize the concentration of the purified GST or GST-tagged protein to be used with Pierce Glutathione Coated Plates?
Yes. We recommend using a solution of the purified GST or GST-tagged protein diluted in Phosphate Buffered Saline (PBS), pH 7.2 in a concentration ranging from 0.01-1 µg/mL. During the coupling, non-ionic detergents like Tween-20 surfactant should not be present, as these will suppress the coupling.
Which type of molecules can I use with Pierce Glutathione Coated Plates?
The Pierce Glutathione Coated Plates are designed for optimal detection of glutathione-S-transferase (GST)-tagged proteins or purified GST.