5-Aminovaleric acid, 97%
5-Aminovaleric acid, 97%
5-Aminovaleric acid, 97%
Thermo Scientific Chemicals

5-Aminovaleric acid, 97%

CAS: 660-88-8 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 153910050
Price (USD)
48.65
Online exclusive
54.20
Save 5.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
48.65
Online exclusive
54.20
Save 5.55
Each
Chemical Identifiers
CAS660-88-8
IUPAC Name5-aminopentanoic acid
Molecular FormulaC5H11NO2
InChI KeyJJMDCOVWQOJGCB-UHFFFAOYSA-N
SMILESNCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Solubility(5 % in water) Clear colorless to light yellow
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with HClO496.0 to 104.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only