Yeast tRNA
Invitrogen™
Yeast tRNA
Yeast tRNA is suitable for use as a carrier in nucleic acid purification and precipitation procedures. It is supplied inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1540102950 mg
1540101125 mg
Catalog number 15401029
Price (USD)
281.65
Online exclusive
310.00
Save 28.35
Each
Quantity:
50 mg
Price (USD)
281.65
Online exclusive
310.00
Save 28.35
Each
Yeast tRNA is suitable for use as a carrier in nucleic acid purification and precipitation procedures. It is supplied in lyophilized form.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Carrier for Nucleic Acid Precipitation
FormLyophilized Powder
Product TypeYeast tRNA
Quantity50 mg
Reagent TypeYeast tRNA
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Citations & References (1)
Citations & References
Abstract
A whole-genome mouse BAC microarray with 1-Mb resolution for analysis of DNA copy number changes by array comparative genomic hybridization.
Authors:Chung YJ, Jonkers J, Kitson H, Fiegler H, Humphray S, Scott C, Hunt S, Yu Y, Nishijima I, Velds A, Holstege H, Carter N, Bradley A,
Journal:Genome Res
PubMed ID:14707179
'Microarray-based comparative genomic hybridization (CGH) has become a powerful method for the genome-wide detection of chromosomal imbalances. Although BAC microarrays have been used for mouse CGH studies, the resolving power of these analyses was limited because high-density whole-genome mouse BAC microarrays were not available. We therefore developed a mouse BAC ... More