UltraPure™ Glycerol, 3.8 L - FAQs
UltraPure™ Glycerol, 3.8 L - FAQs
View additional product information for UltraPure™ Glycerol - FAQs (15514011, 15514029)
1 product FAQs found
What is the storage media for the competent cells?
Preparation procedures and formulations for all of our competent cells are proprietary. All chemically competent cells are delivered in an aqueous solution that contains a mixture of salts, along with a freezing stabilizer such as glycerol or DMSO.