Bovine Serum Albumin - FAQs
Bovine Serum Albumin - FAQs
View additional product information for Bovine Serum Albumin - FAQs (15561020)
1 product FAQs found
What is the Bovine Serum Albumin (BSA) (Cat. No. 15561020) dissolved in?
BSA is supplied sterile in deionized water.