UltraPure™ Salmon Sperm DNA Solution - FAQs
UltraPure™ Salmon Sperm DNA Solution - FAQs
View additional product information for UltraPure™ Salmon Sperm DNA Solution - FAQs (15632011)
3 product FAQs found
What species of salmon is Ultrapure Salmon Sperm DNA derived from?
The species of salmon is Oncerhyncus keta.
Is Ultrapure Salmon Sperm DNA double stranded? How can I make it single stranded?
Ultrapure Salmon Sperm DNA is double stranded. It can be made single stranded by heating to 95 degrees C and then cooling rapidly on ice. Cooling is usually done with the DNA diluted in a hybridization solution which keeps it single stranded because of the large dilution volume.
What concentration should the UltraPure Salmon Sperm DNA Solution (Cat. No. 15632011) be used at?
Carrier DNA is typically used at a concentration of 100 µg/mL in both the prehybridization and hybridization solutions.